missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DGKZ Partial ORF (AAH41770, 786 a.a. - 885 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008525-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16148725

  • 6845.00 SEK / 25µg
Estimated Shipment: 10-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]

Sequence: DAAPPQGEELIEAAKRNDFCKLQELHRAGGDLMHRDEQSRTLLHHAVSTGSKDVVRYLLDHAPPEILDAVEENGETCLHQAAALGQRTICHYIVEAGASL
Specifications

Specifications

AAH41770
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DAAPPQGEELIEAAKRNDFCKLQELHRAGGDLMHRDEQSRTLLHHAVSTGSKDVVRYLLDHAPPEILDAVEENGETCLHQAAALGQRTICHYIVEAGASL
RUO
DGKZ
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8525
DGKZ (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DAGK5/DAGK6/DGK-ZETA/hDGKzeta
DGKZ
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers