missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DGKZ Partial ORF (AAH41770, 786 a.a. - 885 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH41770 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8525 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16138725
|
Abnova™
H00008525-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16148725
|
Abnova™
H00008525-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]
Sequence: DAAPPQGEELIEAAKRNDFCKLQELHRAGGDLMHRDEQSRTLLHHAVSTGSKDVVRYLLDHAPPEILDAVEENGETCLHQAAALGQRTICHYIVEAGASLSpecifications
AAH41770 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DAGK5/DAGK6/DGK-ZETA/hDGKzeta | |
DGKZ | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8525 | |
DGKZ (Human) Recombinant Protein (Q01) | |
DAAPPQGEELIEAAKRNDFCKLQELHRAGGDLMHRDEQSRTLLHHAVSTGSKDVVRYLLDHAPPEILDAVEENGETCLHQAAALGQRTICHYIVEAGASL | |
RUO | |
DGKZ | |
Wheat Germ (in vitro) | |
GST | |
Liquid |