missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DGUOK Partial ORF (NP_001920, 1 a.a. - 89 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001716-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16117184

  • 6845.00 SEK / 25µg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

In mammalian cells, the phosphorylation of purine deoxyribonucleosides is mediated predominantly by two deoxyribonucleoside kinases, cytosolic deoxycytidine kinase and mitochondrial deoxyguanosine kinase. The protein encoded by this gene is responsible for phosphorylation of purine deoxyribonucleosides in the mitochondrial matrix. In addition, this protein phosphorylates several purine deoxyribonucleoside analogs used in the treatment of lymphoproliferative disorders, and this phosphorylation is critical for the effectiveness of the analogs. Alternative splice variants encoding different protein isoforms have been described for this gene. [provided by RefSeq]

Sequence: MAAGRLFLSRLRAPFSSMAKSPLEGVSSSRGLHAGRGPRRLSIEGNIGLHCPKSWKLAGYDVPGASTMVLHIPDIFLFEPPESTAGALP
Specifications

Specifications

NP_001920
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.53kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAGRLFLSRLRAPFSSMAKSPLEGVSSSRGLHAGRGPRRLSIEGNIGLHCPKSWKLAGYDVPGASTMVLHIPDIFLFEPPESTAGALP
RUO
DGUOK
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1716
DGUOK (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
dGK
DGUOK
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers