Learn More
Abnova™ Human DUSP9 Full-length ORF (AAH60837.1, 1 a.a. - 384 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product shows selectivity for members of the ERK family of MAP kinases, is expressed only in placenta, kidney, and fetal liver, and is localized to the cytoplasm and nucleus. [provided by RefSeq]
Specifications
Specifications
Accession Number | AAH60837.1 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1852 |
Molecular Weight (g/mol) | 68.3kDa |
Name | DUSP9 (Human) Recombinant Protein (P01) |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 ug |
Immunogen | MEGLGRSCLWLRRELSPPRPRLLLLDCRSRELYESARIGGALSVALPALLLRRLRRGSLSVRALLPGPPLQPPPPAPVLLYDQGGGRRRRGEAEAEAEEWEAESVLGTLLQKLREEGYLAYYLQGGFSRFQAECPHLCETSLAGRAGSSMAPLPGPVPVVGLGSLCLGSDCSDAESEADRDSMSCGLDSEGATPPPVGLRASFPVQILPNLYLGSARDSANLESLAKLGIRYILNVTPNLPNFFEKNGDFHYKQIPISDHWSQNLSRFFPEAIEFIDEALSQNRGVLVHCLAGVSRSVTVTVAYLMQKLHLSLNDAYDLVKRKKSNISPNFNFMGQLLDFERSLRLEERHSQEQGSGGQASAASNPPSFFTTPTSDGAFELAPT |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.