missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ENAH Partial ORF (NP_001008493.1, 291 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055740-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
Sequence: LQAASQPAETPSQQGIVLGPLAPPPPPPLPPGPAQASVALPPPPGPPPPPPLPSTGPPPPPPPPPLPNQVPPPPPPPPAPPLPASGFFLASMSEDNRPLTSpecifications
NP_001008493.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LQAASQPAETPSQQGIVLGPLAPPPPPPLPPGPAQASVALPPPPGPPPPPPLPSTGPPPPPPPPPLPNQVPPPPPPPPAPPLPASGFFLASMSEDNRPLT | |
RUO | |
ENAH | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
55740 | |
ENAH (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ENA|MENA|NDPP1 | |
ENAH | |
Yes |