Learn More
Abnova™ Human ENAM Partial ORF (NP_114095, 1043 a.a. - 1141 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
Dental enamel is a highly mineralized tissue with 85% of its volume occupied by unusually large, highly organized, hydroxyapatite crystals. This highly organized and unusual structure is thought to be rigorously controlled in ameloblasts through the interaction of a number of organic matrix molecules that include enamelin, amelogenin (AMELX; MIM 300391), ameloblastin (AMBN; MIM 601259), tuftelin (TUFT1; MIM 600087), dentine sialophosphoprotein (DSPP; MIM 125485), and a variety of enzymes. Enamelin is the largest protein in the enamel matrix of developing teeth and comprises approximately 5% of total enamel matrix protein.[supplied by OMIM]
Specifications
Specifications
Accession Number | NP_114095 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10117 |
Molecular Weight (g/mol) | 36.63kDa |
Name | ENAM (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 μg |
Immunogen | ERQQQRPSNILHLPCFGSKLAKHHSSTTGTPSSDGRQSPFDGDSITPTENPNTLVELATEEQFKSINVDPLDADEHSPFEFLQRGTNVQDQVQDCLLLQ |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.