missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ENC1 Full-length ORF (NP_003624.1, 1 a.a. - 589 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008507-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122512

  • 4055.00 SEK / 10µg
Estimated Shipment: 14-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DNA damage and/or hyperproliferative signals activate wildtype p53 tumor suppressor protein (TP53; MIM 191170), inducing cell cycle arrest or apoptosis. Mutations that inactivate p53 occur in 50% of all tumors. Polyak et al. (1997) [PubMed 9305847] used serial analysis of gene expression (SAGE) to evaluate cellular mRNA levels in a colorectal cancer cell line transfected with p53. Of 7,202 transcripts identified, only 14 were expressed at levels more than 10-fold higher in p53-expressing cells than in control cells. Polyak et al. (1997) [PubMed 9305847] termed these genes 'p53-induced genes,' or PIGs, several of which were predicted to encode redox-controlling proteins. They noted that reactive oxygen species (ROS) are potent inducers of apoptosis. Flow cytometric analysis showed that p53 expression induces ROS production, which increases as apoptosis progresses under some conditions. The authors stated that the PIG10 gene, also called ENC1, encodes an actin-binding protein.[supplied by OMIM]

Sequence: MSVSVHENRKSRASSGSINIYLFHKSSYADSVLTHLNLLRQQRLFTDVLLHAGNRTFPCHRAVLAACSRYFEAMFSGGLKESQDSEVNFDNSIHPEVLELLLDYAYSSRVIINEENAESLLEAGDMLEFQDIRDACAEFLEKNLHPTNCLGMLLLSDAHQCTKLYELSWRMCLSNFQTIRKNEDFLQLPQDMVVQLLSSEELETEDERLVYESAINWISYDLKKRYCYLPELLQTVRLALLPAIYLMENVAMEELITKQRKSKEIVEEAIRCKLKILQNDGVVTSLCARPRKTGHALFLLGGQTFMCDKLYLVDQKAKEIIPKADIPSPRKEFSACAIGCKVYITGGRGSENGVSKDVWVYDTLHEEWSKAAPMLVARFGHGSAELKHCLYVVGGHTAATGCLPASPSVSLKQVEHYDPTINKWTMVAPLREGVSNAAVVSAKLKLFAFGGTSVSHDKLPKVQCYDQCENRWTVPATCPQPWRYTAAAVLGNQIFIMGGDTEFSACSAYKFNSETYQWTKVGDVTAKRMSCHAVASGNKLYVVGGYFGIQRCKTLDCYDPTLDVWNSITTVPYSLIPTAFVSTWKHLPS
Specifications

Specifications

NP_003624.1
Liquid
8507
ENC1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSVSVHENRKSRASSGSINIYLFHKSSYADSVLTHLNLLRQQRLFTDVLLHAGNRTFPCHRAVLAACSRYFEAMFSGGLKESQDSEVNFDNSIHPEVLELLLDYAYSSRVIINEENAESLLEAGDMLEFQDIRDACAEFLEKNLHPTNCLGMLLLSDAHQCTKLYELSWRMCLSNFQTIRKNEDFLQLPQDMVVQLLSSEELETEDERLVYESAINWISYDLKKRYCYLPELLQTVRLALLPAIYLMENVAMEELITKQRKSKEIVEEAIRCKLKILQNDGVVTSLCARPRKTGHALFLLGGQTFMCDKLYLVDQKAKEIIPKADIPSPRKEFSACAIGCKVYITGGRGSENGVSKDVWVYDTLHEEWSKAAPMLVARFGHGSAELKHCLYVVGGHTAATGCLPASPSVSLKQVEHYDPTINKWTMVAPLREGVSNAAVVSAKLKLFAFGGTSVSHDKLPKVQCYDQCENRWTVPATCPQPWRYTAAAVLGNQIFIMGGDTEFSACSAYKFNSETYQWTKVGDVTAKRMSCHAVASGNKLYVVGGYFGIQRCKTLDCYDPTLDVWNSITTVPYSLIPTAFVSTWKHLPS
RUO
ENC1
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
92.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CCL28/ENC-1/FLJ39259/KLHL35/KLHL37/NRPB/PIG10/TP53I10
ENC1
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers