Läs mer
Human Endoglin/CD105 PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 26 publications
Brand: R&D Systems FAB10971P-025
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Flow Cytometry.
Specifikationer
| Endoglin/CD105 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, ENG, HHT1FLJ41744, ORW, ORW1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2022 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 166707 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q5T9B9 | |
| ENG | |
| Mouse myeloma cell line NS0-derived recombinant human Endoglin/CD105 Glu26-Gly586 Accession # Q5T9B9 | |
| 25 Tests | |
| Primary | |
| Detects human Endoglin/CD105. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |