Learn More
Abnova™ Human EVC Full-length ORF (AAH85608.1, 1 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002121-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein containing a leucine zipper and a transmembrane domain. This gene has been implicated in both Ellis-van Creveld syndrome (EvC) and Weyers acrodental dysostosis. [provided by RefSeq]
Sequence: MARGGAACKSDARLLLGRDALRPAPALLAPAVLLGAALGLGLGLWLGCRAGRQRTRHQKDDTQNLLKNLESNAPTPSETGSPSRRRKREVQMSKDKEAVDECEPPSNSNITAFALKAKVIYPINQKFRPLADGSSNPSLHENLKQAVLPHQPVEASPSSSLGSLSQGEKDDCSSSSSVHSATSDDRFLSRTFLRVNAFPEVLACESVDVDLCIYSLHLKDLLHLDTALRQEKHMMFIQIFKMCLLDLLPKKKSDDELHQKILSKQEKDLEELEKGLQVKLSNTEMSGAGDSEYITLADVEKKEREYSEQLIDNMEAFWKQMANIQHFLVDQFKCSSSKARQLMMTLTERMIAAEGLLCDSQELQALDALERTMGRAHMAKVIEFLKLQVQEETRCRLAAISHGLELLAGEGKLSGRQKEELLTQQHKAFWQEAERFSREFVQRGKDLVKASLAHQVEGTAKLTLAQEEEQRSFLAEAQPTADPEKFLEAFHEVLERQRLMQCDLEEEENVRATEAVVALCQKAMRRHRDTGKEAVSpecifications
AAH85608.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
86.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DWF-1/EVC1/EVCL/MGC105107 | |
EVC | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2121 | |
EVC (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MARGGAACKSDARLLLGRDALRPAPALLAPAVLLGAALGLGLGLWLGCRAGRQRTRHQKDDTQNLLKNLESNAPTPSETGSPSRRRKREVQMSKDKEAVDECEPPSNSNITAFALKAKVIYPINQKFRPLADGSSNPSLHENLKQAVLPHQPVEASPSSSLGSLSQGEKDDCSSSSSVHSATSDDRFLSRTFLRVNAFPEVLACESVDVDLCIYSLHLKDLLHLDTALRQEKHMMFIQIFKMCLLDLLPKKKSDDELHQKILSKQEKDLEELEKGLQVKLSNTEMSGAGDSEYITLADVEKKEREYSEQLIDNMEAFWKQMANIQHFLVDQFKCSSSKARQLMMTLTERMIAAEGLLCDSQELQALDALERTMGRAHMAKVIEFLKLQVQEETRCRLAAISHGLELLAGEGKLSGRQKEELLTQQHKAFWQEAERFSREFVQRGKDLVKASLAHQVEGTAKLTLAQEEEQRSFLAEAQPTADPEKFLEAFHEVLERQRLMQCDLEEEENVRATEAVVALCQKAMRRHRDTGKEAV | |
RUO | |
EVC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |