missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EVC Full-length ORF (AAH85608.1, 1 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH85608.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2121 |
Molecular Weight (g/mol) | 86.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16184711
|
Abnova™
H00002121-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16174711
|
Abnova™
H00002121-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein containing a leucine zipper and a transmembrane domain. This gene has been implicated in both Ellis-van Creveld syndrome (EvC) and Weyers acrodental dysostosis. [provided by RefSeq]
Sequence: MARGGAACKSDARLLLGRDALRPAPALLAPAVLLGAALGLGLGLWLGCRAGRQRTRHQKDDTQNLLKNLESNAPTPSETGSPSRRRKREVQMSKDKEAVDECEPPSNSNITAFALKAKVIYPINQKFRPLADGSSNPSLHENLKQAVLPHQPVEASPSSSLGSLSQGEKDDCSSSSSVHSATSDDRFLSRTFLRVNAFPEVLACESVDVDLCIYSLHLKDLLHLDTALRQEKHMMFIQIFKMCLLDLLPKKKSDDELHQKILSKQEKDLEELEKGLQVKLSNTEMSGAGDSEYITLADVEKKEREYSEQLIDNMEAFWKQMANIQHFLVDQFKCSSSKARQLMMTLTERMIAAEGLLCDSQELQALDALERTMGRAHMAKVIEFLKLQVQEETRCRLAAISHGLELLAGEGKLSGRQKEELLTQQHKAFWQEAERFSREFVQRGKDLVKASLAHQVEGTAKLTLAQEEEQRSFLAEAQPTADPEKFLEAFHEVLERQRLMQCDLEEEENVRATEAVVALCQKAMRRHRDTGKEAVSpecifications
AAH85608.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
86.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MARGGAACKSDARLLLGRDALRPAPALLAPAVLLGAALGLGLGLWLGCRAGRQRTRHQKDDTQNLLKNLESNAPTPSETGSPSRRRKREVQMSKDKEAVDECEPPSNSNITAFALKAKVIYPINQKFRPLADGSSNPSLHENLKQAVLPHQPVEASPSSSLGSLSQGEKDDCSSSSSVHSATSDDRFLSRTFLRVNAFPEVLACESVDVDLCIYSLHLKDLLHLDTALRQEKHMMFIQIFKMCLLDLLPKKKSDDELHQKILSKQEKDLEELEKGLQVKLSNTEMSGAGDSEYITLADVEKKEREYSEQLIDNMEAFWKQMANIQHFLVDQFKCSSSKARQLMMTLTERMIAAEGLLCDSQELQALDALERTMGRAHMAKVIEFLKLQVQEETRCRLAAISHGLELLAGEGKLSGRQKEELLTQQHKAFWQEAERFSREFVQRGKDLVKASLAHQVEGTAKLTLAQEEEQRSFLAEAQPTADPEKFLEAFHEVLERQRLMQCDLEEEENVRATEAVVALCQKAMRRHRDTGKEAV | |
RUO | |
EVC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
2121 | |
EVC (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DWF-1/EVC1/EVCL/MGC105107 | |
EVC | |
Recombinant | |
wheat germ expression system |