missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EYA3 Partial ORF (NP_001981.2, 272 a.a. - 374 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
Description
This gene encodes a member of the eyes absent (EYA) family of proteins. The encoded protein may act as a transcriptional activator and have a role during development. A similar protein in mice acts as a transcriptional activator. [provided by RefSeq]
Sequence: KDTDDQSRKNMTSKNRGKRKADATSSQDSELERVFLWDLDETIIIFHSLLTGSYAQKYGKDPTVVIGSGLTMEEMIFEVADTHLFFNDLEECDQVHVEDVASD
Specifications
Specifications
Accession Number | NP_001981.2 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2140 |
Molecular Weight (g/mol) | 37.07kDa |
Name | EYA3 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 ug |
Immunogen | KDTDDQSRKNMTSKNRGKRKADATSSQDSELERVFLWDLDETIIIFHSLLTGSYAQKYGKDPTVVIGSGLTMEEMIFEVADTHLFFNDLEECDQVHVEDVASD |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human EYA3 Partial ORF (NP_001981.2, 272 a.a. - 374 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction