missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FAM55A Full-length ORF (NP_689528.1, 1 a.a. - 405 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4630.00 SEK - 7020.00 SEK

Specifications

Accession Number NP_689528.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 120400
Molecular Weight (g/mol) 73.3kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16197473
View Documents
Abnova™
H00120400-P01.25ug
25 ÎĽg
7020.00 SEK
25µg
Please to purchase this item. Need a web account? Register with us today!
16187473
View Documents
Abnova™
H00120400-P01.10ug
10 ÎĽg
4630.00 SEK
10µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence: MSSPALTAGASGKVMDFNNGTYLVSFTLFWEGQVSLSLLLIHPSEGASALWRARNQGYDKIIFKGKFVNGTSHVFTECGLTLNSNAELCEYLDDRDQEAFYCMKPQHMPCEALTYMTTRNREVSYLTDKENSLFHRSKVGVEMMKDRKHIDVTNCNKREKIEETCQVGMKPPVPGGYTLQGKWITTFCNQVQLDTIKINGCLKGKLIYLLGDSTLRQWIYYFPKVVKTLKFFDLHETGIFKKHLLLDAERHTQIQWKKHSYPFVTFQLYSLIDHDYIPREIDRLSGDKNTAIVITFGQHFRPFPIDIFIRRAIGVQKAIERLFLRSPATKVIIKTENIREMHIETERFVDFHGYIHYLIMKDIFKDLNVGIIDAWDMTIAYGTDTIHPPDHVIGNQINMFLNYIC
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human FAM55A Full-length ORF (NP_689528.1, 1 a.a. - 405 a.a.) Recombinant Protein with GST-tag at N-terminal

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.