missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FAM9B Partial ORF (NP_995321.1, 31 a.a. - 86 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00171483-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: DVTDEHGEREPFAETDEHTGANTKKPEDTAEDLTAKRKRMKMDKTCSKTKNKSKHASpecifications
NP_995321.1 | |
Liquid | |
171483 | |
FAM9B (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ40182 | |
FAM9B | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.9kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DVTDEHGEREPFAETDEHTGANTKKPEDTAEDLTAKRKRMKMDKTCSKTKNKSKHA | |
RUO | |
FAM9B | |
Wheat Germ (in vitro) | |
GST |