missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FARP1 Partial ORF (NP_005757.1, 471 a.a. - 549 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010160-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16172006

  • 4515.00 SEK / 10µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene was originally isolated through subtractive hybridization due to its increased expression in differentiated chondrocytes versus dedifferentiated chondrocytes. The resulting protein contains a predicted ezrin-like domain, a Dbl homology domain, and a pleckstrin homology domain. It is believed to be a member of the band 4.1 superfamily whose members link the cytoskeleton to the cell membrane. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq]

Sequence: TGSLTGSPHLSELSVNSQGGVAPANVTLSPNLSPDTKQASPLISPLLNDQACPRTDDEDEGRRKRFPTDKAYFIAKEVS
Specifications

Specifications

NP_005757.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.43kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TGSLTGSPHLSELSVNSQGGVAPANVTLSPNLSPDTKQASPLISPLLNDQACPRTDDEDEGRRKRFPTDKAYFIAKEVS
RUO
FARP1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10160
FARP1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CDEP/MGC87400/PLEKHC2
FARP1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers