missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FARP1 Partial ORF (NP_005757.1, 471 a.a. - 549 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_005757.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10160 |
Molecular Weight (g/mol) | 34.43kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16182006
|
Abnova™
H00010160-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16172006
|
Abnova™
H00010160-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene was originally isolated through subtractive hybridization due to its increased expression in differentiated chondrocytes versus dedifferentiated chondrocytes. The resulting protein contains a predicted ezrin-like domain, a Dbl homology domain, and a pleckstrin homology domain. It is believed to be a member of the band 4.1 superfamily whose members link the cytoskeleton to the cell membrane. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq]
Sequence: TGSLTGSPHLSELSVNSQGGVAPANVTLSPNLSPDTKQASPLISPLLNDQACPRTDDEDEGRRKRFPTDKAYFIAKEVSSpecifications
NP_005757.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.43kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDEP/MGC87400/PLEKHC2 | |
FARP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10160 | |
FARP1 (Human) Recombinant Protein (Q01) | |
TGSLTGSPHLSELSVNSQGGVAPANVTLSPNLSPDTKQASPLISPLLNDQACPRTDDEDEGRRKRFPTDKAYFIAKEVS | |
RUO | |
FARP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |