missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human FCRL1 Partial ORF (NP_443170, 23 a.a. - 117 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16181477

Abnova™ Human FCRL1 Partial ORF (NP_443170, 23 a.a. - 117 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16181477
25 μg, 25µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16181477

Brand: Abnova™ H00115350Q01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein contains three extracellular C2-like immunoglobulin domains, a transmembrane domain and a cytoplasmic domain with two immunoreceptor-tyrosine activation motifs. This protein may play a role in the regulation of cancer cell growth. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: ASPSHPTEGSPVTLTCKMPFLQSSDAQFQFCFFRDTRALGPGWSSSPKLQIAAMWKEDTGSYWCEAQTMASKVLRSRRSQINVHRVPVADVSLET

Specifications

Accession Number NP_443170
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 115350
Molecular Weight (g/mol) 36.19kDa
Name FCRL1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen ASPSHPTEGSPVTLTCKMPFLQSSDAQFQFCFFRDTRALGPGWSSSPKLQIAAMWKEDTGSYWCEAQTMASKVLRSRRSQINVHRVPVADVSLET
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp667O1421/FCRH1/IFGP1/IRTA5/RP11-367J7.7
Common Name FCRL1
Gene Symbol FCRL1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human FCRL1 Partial ORF (NP_443170, 23 a.a. - 117 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.