missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FOXA3 Partial ORF (NP_004488.2, 266 a.a. - 350 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003171-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139444

  • 6845.00 SEK / 25µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. The crystal structure of a similar protein in rat has been resolved. [provided by RefSeq]

Sequence: EDVGALDCGSPASSTPYFTGLELPGELKLDAPYNFNHPFSINNLMSEQTPAPPKLDVGFGGYGAEGGEPGVYYQGLYSRSLLNAS
Specifications

Specifications

NP_004488.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.09kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EDVGALDCGSPASSTPYFTGLELPGELKLDAPYNFNHPFSINNLMSEQTPAPPKLDVGFGGYGAEGGEPGVYYQGLYSRSLLNAS
RUO
FOXA3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3171
FOXA3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FKHH3/HNF3G/MGC10179/TCF3G
FOXA3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers