missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FZD4 Full-length ORF (NP_036325.2, 1 a.a. - 537 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008322-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16162362

  • 4515.00 SEK / 10µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. This protein may play a role as a positive regulator of the Wingless type MMTV integration site signaling pathway. A transcript variant retaining intronic sequence and encoding a shorter isoform has been described, however, its expression is not supported by other experimental evidence. [provided by RefSeq]

Sequence: MAWRGAGPSVPGAPGGVGLSLGLLLQLLLLLGPARGFGDEEERRCDPIRISMCQNLGYNVTKMPNLVGHELQTDAELQLTTFTPLIQYGCSSQLQFFLCSVYVPMCTEKINIPIGPCGGMCLSVKRRCEPVLKEFGFAWPESLNCSKFPPQNDHNHMCMEGPGDEEVPLPHKTPIQPGEECHSVGTNSDQYIWVKRSLNCVLKCGYDAGLYSRSAKEFTDIWMAVWASLCFISTAFTVLTFLIDSSRFSYPERPIIFLSMCYNIYSIAYIVRLTVGRERISCDFEEAAEPVLIQEGLKNTGCAIIFLLMYFFGMASSIWWVILTLTWFLAAGLKWGHEAIEMHSSYFHIAAWAIPAVKTIVILIMRLVDADELTGLCYVGNQNLDALTGFVVAPLFTYLVIGTLFIAAGLVALFKIRSNLQKDGTKTDKLERLMVKIGVFSVLYTVPATCVIACYFYEISNWALFRYSADDSNMAVEMLKIFMSLLVGITSGMWIWSAKTLHTWQKCSNRLVNSGKVKREKRGNGWVKPGKGSETVV
Specifications

Specifications

NP_036325.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
86.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD344/EVR1/FEVR/FZD4S/Fz-4/FzE4/GPCR/MGC34390
FZD4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8322
FZD4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAWRGAGPSVPGAPGGVGLSLGLLLQLLLLLGPARGFGDEEERRCDPIRISMCQNLGYNVTKMPNLVGHELQTDAELQLTTFTPLIQYGCSSQLQFFLCSVYVPMCTEKINIPIGPCGGMCLSVKRRCEPVLKEFGFAWPESLNCSKFPPQNDHNHMCMEGPGDEEVPLPHKTPIQPGEECHSVGTNSDQYIWVKRSLNCVLKCGYDAGLYSRSAKEFTDIWMAVWASLCFISTAFTVLTFLIDSSRFSYPERPIIFLSMCYNIYSIAYIVRLTVGRERISCDFEEAAEPVLIQEGLKNTGCAIIFLLMYFFGMASSIWWVILTLTWFLAAGLKWGHEAIEMHSSYFHIAAWAIPAVKTIVILIMRLVDADELTGLCYVGNQNLDALTGFVVAPLFTYLVIGTLFIAAGLVALFKIRSNLQKDGTKTDKLERLMVKIGVFSVLYTVPATCVIACYFYEISNWALFRYSADDSNMAVEMLKIFMSLLVGITSGMWIWSAKTLHTWQKCSNRLVNSGKVKREKRGNGWVKPGKGSETVV
RUO
FZD4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers