Learn More
Abnova™ Human FZD4 Full-length ORF (NP_036325.2, 1 a.a. - 537 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_036325.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8322 |
Molecular Weight (g/mol) | 86.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16172362
|
Abnova™
H00008322-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16162362
|
Abnova™
H00008322-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. This protein may play a role as a positive regulator of the Wingless type MMTV integration site signaling pathway. A transcript variant retaining intronic sequence and encoding a shorter isoform has been described, however, its expression is not supported by other experimental evidence. [provided by RefSeq]
Sequence: MAWRGAGPSVPGAPGGVGLSLGLLLQLLLLLGPARGFGDEEERRCDPIRISMCQNLGYNVTKMPNLVGHELQTDAELQLTTFTPLIQYGCSSQLQFFLCSVYVPMCTEKINIPIGPCGGMCLSVKRRCEPVLKEFGFAWPESLNCSKFPPQNDHNHMCMEGPGDEEVPLPHKTPIQPGEECHSVGTNSDQYIWVKRSLNCVLKCGYDAGLYSRSAKEFTDIWMAVWASLCFISTAFTVLTFLIDSSRFSYPERPIIFLSMCYNIYSIAYIVRLTVGRERISCDFEEAAEPVLIQEGLKNTGCAIIFLLMYFFGMASSIWWVILTLTWFLAAGLKWGHEAIEMHSSYFHIAAWAIPAVKTIVILIMRLVDADELTGLCYVGNQNLDALTGFVVAPLFTYLVIGTLFIAAGLVALFKIRSNLQKDGTKTDKLERLMVKIGVFSVLYTVPATCVIACYFYEISNWALFRYSADDSNMAVEMLKIFMSLLVGITSGMWIWSAKTLHTWQKCSNRLVNSGKVKREKRGNGWVKPGKGSETVVSpecifications
NP_036325.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
86.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAWRGAGPSVPGAPGGVGLSLGLLLQLLLLLGPARGFGDEEERRCDPIRISMCQNLGYNVTKMPNLVGHELQTDAELQLTTFTPLIQYGCSSQLQFFLCSVYVPMCTEKINIPIGPCGGMCLSVKRRCEPVLKEFGFAWPESLNCSKFPPQNDHNHMCMEGPGDEEVPLPHKTPIQPGEECHSVGTNSDQYIWVKRSLNCVLKCGYDAGLYSRSAKEFTDIWMAVWASLCFISTAFTVLTFLIDSSRFSYPERPIIFLSMCYNIYSIAYIVRLTVGRERISCDFEEAAEPVLIQEGLKNTGCAIIFLLMYFFGMASSIWWVILTLTWFLAAGLKWGHEAIEMHSSYFHIAAWAIPAVKTIVILIMRLVDADELTGLCYVGNQNLDALTGFVVAPLFTYLVIGTLFIAAGLVALFKIRSNLQKDGTKTDKLERLMVKIGVFSVLYTVPATCVIACYFYEISNWALFRYSADDSNMAVEMLKIFMSLLVGITSGMWIWSAKTLHTWQKCSNRLVNSGKVKREKRGNGWVKPGKGSETVV | |
RUO | |
FZD4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
8322 | |
FZD4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD344/EVR1/FEVR/FZD4S/Fz-4/FzE4/GPCR/MGC34390 | |
FZD4 | |
Recombinant | |
wheat germ expression system |