missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human G10 Partial ORF (NP_003901, 35 a.a. - 144 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
Sequence: EPHEGKRKVESLWPIFRIHHQKTRYIFDLFYKRKAISRELYEYCIKEGYADKNLIAKWKKQGYENLCCLRCIQTRDTNFGTNCICRVPKSKLEVGRIIECTHCGCRGCSG
Specifications
Specifications
Accession Number | NP_003901 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8896 |
Molecular Weight (g/mol) | 37.84kDa |
Name | G10 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 μg |
Immunogen | EPHEGKRKVESLWPIFRIHHQKTRYIFDLFYKRKAISRELYEYCIKEGYADKNLIAKWKKQGYENLCCLRCIQTRDTNFGTNCICRVPKSKLEVGRIIECTHCGCRGCSG |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human G10 Partial ORF (NP_003901, 35 a.a. - 144 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction