missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human GALNTL5 Full-length ORF (NP_660335.1, 1 a.a. - 443 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16139083

Abnova™ Human GALNTL5 Full-length ORF (NP_660335.1, 1 a.a. - 443 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16139083
25 μg, 25µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16139083

Brand: Abnova™ H00168391P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

Sequence: MRNAIIQGLFYGSLTFGIWTALLFIYLHHNHVSSWQKKSQEPLSAWSPGKKVHQQIIYGSEQIPKPHVIVKRTDEDKAKSMLGTDFNHTNPELHKELLKYGFNVIISRSLGIEREVPDTRSKMRLQKHYPARLPTASIVICFYNEECNALFQTMSSVTNLTPHYFLEEIILVDDMSKVDDLKEKLDYHLETFRGKVKIIRNKKREGLIRARLIGASHASGDVLVFLDSHCEVNRVWLEPLLHAIAKDPKMVVCPLIDVIDDRTLEYKPSPLVRGTFDWNLQFKWDNVFSYEMDGPEGSTKPIRSPAMSGGIFAIRRHYFNEIGQYDKDMDFWGRENLELSLRIWMCGGQLFIIPCSRVGHISKKQTGKPSTIISAMTHNYLRLVHVWLDEYKEQFFLRKPGLKYVTYGNIRERVELRKRLGCKSFQWYLDNVFPELEASVNSL

Specifications

Accession Number NP_660335.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 168391
Molecular Weight (g/mol) 77.9kDa
Name GALNTL5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MRNAIIQGLFYGSLTFGIWTALLFIYLHHNHVSSWQKKSQEPLSAWSPGKKVHQQIIYGSEQIPKPHVIVKRTDEDKAKSMLGTDFNHTNPELHKELLKYGFNVIISRSLGIEREVPDTRSKMRLQKHYPARLPTASIVICFYNEECNALFQTMSSVTNLTPHYFLEEIILVDDMSKVDDLKEKLDYHLETFRGKVKIIRNKKREGLIRARLIGASHASGDVLVFLDSHCEVNRVWLEPLLHAIAKDPKMVVCPLIDVIDDRTLEYKPSPLVRGTFDWNLQFKWDNVFSYEMDGPEGSTKPIRSPAMSGGIFAIRRHYFNEIGQYDKDMDFWGRENLELSLRIWMCGGQLFIIPCSRVGHISKKQTGKPSTIISAMTHNYLRLVHVWLDEYKEQFFLRKPGLKYVTYGNIRERVELRKRLGCKSFQWYLDNVFPELEASVNSL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GALNT15
Common Name GALNTL5
Gene Symbol GALNTL5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human GALNTL5 Full-length ORF (NP_660335.1, 1 a.a. - 443 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.