missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human GAS2L1 Full-length ORF (NP_006469.2, 1 a.a. - 681 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16135462

Abnova™ Human GAS2L1 Full-length ORF (NP_006469.2, 1 a.a. - 681 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16135462
10 μg, 10µg
missing translation for 'orderingAttributeHoverText'
Quantity:
10 μg
25 μg
missing translation for 'unitSize'
10µg
25µg
This item is not returnable. View return policy

Product Code. 16135462

Brand: Abnova™ H00010634P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene, a member of the GAS2 family, is similar in sequence to the mouse protein Gas2, an actin-associated protein expressed at high levels in growth-arrested cells. Expression of the mouse Gas2 gene is negatively regulated by serum and growth factors. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MADPVAGIAGSAAKSVRPFRSSEAYVEAMKEDLAEWLNALYGLGLPGGGDGFLTGLATGTTLCQHANAVTEAARALAAARPARGVAFQAHSVVPGSFMARDNVATFIGWCRVELGVPEVLMFETEDLVLRKNEKSVVLCLLEVARRGARLGLLAPRLVQFEQEIERELRAAPPAPNAPAAGEDTTETAPAPGTPARGPRMTPSDLRNLDELVREILGRCTCPDQFPMIKVSEGKYRVGDSSLLIFVRVLRSHVMVRVGGGWDTLEHYLDKHDPCRCSSTAHRPPQPRVCTFSPQRVSPTTSPRPASPVPGSERRGSRPEMTPVSLRSTKEGPETPPRPRDQLPPHPRSRRYSGDSDSSASSAQSGPLGTRSDDTGTGPRRERPSRRLTTGTPASPRRPPALRSQSRDRLDRGRPRGAPGGRGAQLSVPSPARRARSQSREEQAVLLVRRDRDGQHSWVPRGRGSGGSGRSTPQTPRARSPAAPRLSRVSSPSPELGTTPASIFRTPLQLDPQQEQQLFRRLEEEFLANARALEAVASVTPTGPAPDPARAPDPPAPDSAYCSSSSSSSSLSVLGGKCGQPGDSGRTANGLPGPRSQALSSSSDEGSPCPGMGGPLDAPGSPLACTEPSRTWARGRMDTQPDRKPSRIPTPRGPRRPSGPAELGTWHALHSVTPRAEPDSWM

Specifications

Accession Number NP_006469.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10634
Molecular Weight (g/mol) 99.1kDa
Name GAS2L1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MADPVAGIAGSAAKSVRPFRSSEAYVEAMKEDLAEWLNALYGLGLPGGGDGFLTGLATGTTLCQHANAVTEAARALAAARPARGVAFQAHSVVPGSFMARDNVATFIGWCRVELGVPEVLMFETEDLVLRKNEKSVVLCLLEVARRGARLGLLAPRLVQFEQEIERELRAAPPAPNAPAAGEDTTETAPAPGTPARGPRMTPSDLRNLDELVREILGRCTCPDQFPMIKVSEGKYRVGDSSLLIFVRVLRSHVMVRVGGGWDTLEHYLDKHDPCRCSSTAHRPPQPRVCTFSPQRVSPTTSPRPASPVPGSERRGSRPEMTPVSLRSTKEGPETPPRPRDQLPPHPRSRRYSGDSDSSASSAQSGPLGTRSDDTGTGPRRERPSRRLTTGTPASPRRPPALRSQSRDRLDRGRPRGAPGGRGAQLSVPSPARRARSQSREEQAVLLVRRDRDGQHSWVPRGRGSGGSGRSTPQTPRARSPAAPRLSRVSSPSPELGTTPASIFRTPLQLDPQQEQQLFRRLEEEFLANARALEAVASVTPTGPAPDPARAPDPPAPDSAYCSSSSSSSSLSVLGGKCGQPGDSGRTANGLPGPRSQALSSSSDEGSPCPGMGGPLDAPGSPLACTEPSRTWARGRMDTQPDRKPSRIPTPRGPRRPSGPAELGTWHALHSVTPRAEPDSWM
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GAR22/MGC17243
Common Name GAS2L1
Gene Symbol GAS2L1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human GAS2L1 Full-length ORF (NP_006469.2, 1 a.a. - 681 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.