Learn More
Abnova™ Human GBP6 Full-length ORF (NP_940862.1, 1 a.a. - 633 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00163351-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Guanylate-binding proteins, such as GBP6, are induced by interferon and hydrolyze GTP to both GDP and GMP (Olszewski et al., 2006 [PubMed 16689661]).[supplied by OMIM]
Sequence: MESGPKMLAPVCLVENNNEQLLVNQQAIQILEKISQPVVVVAIVGLYRTGKSYLMNHLAGQNHGFPLGSTVQSETKGIWMWCVPHPSKPNHTLVLLDTEGLGDVEKGDPKNDSWIFALAVLLCSTFVYNSMSTINHQALEQLHYVTELTELIKAKSSPRPDGVEDSTEFVSFFPDFLWTVRDFTLELKLNGHPITEDEYLENALKLIQGNNPRVQTSNFPRECIRRFFPKRKCFVFDRPTNDKDLLANIEKVSEKQLDPKFQEQTNIFCSYIFTHARTKTLREGITVTGNRLGTLAVTYVEAINSGAVPCLENAVITLAQRENSAAVQRASDYYSQQMAQRVKFPTDTLQELLDVHAACEREAIAIFMEHSFKDENQEFQKKFMETTMNKKGDFLLQNEESSVQYCQAKLNELSKGLMESISAGSFSVPGGHKLYMETKERIEQDYWQVPRKGVKAKEVFQRFLESQMVIEESILQSDKALTDREKAVAVDRAKKEAAEKEQELLKQKLQEQQQQMEAQVKSRKENIAQLKEKLQMEREHLLREQIMMLEHTQKVQNDWLHEGFKKKYEEMNAEISQFKRMIDTTKNDDTPWIARTLDNLADELTAILSAPAKLIGHGVKGVSSLFKKHKLPFSpecifications
NP_940862.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
98.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686G0786/FLJ39135 | |
GBP6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
163351 | |
GBP6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MESGPKMLAPVCLVENNNEQLLVNQQAIQILEKISQPVVVVAIVGLYRTGKSYLMNHLAGQNHGFPLGSTVQSETKGIWMWCVPHPSKPNHTLVLLDTEGLGDVEKGDPKNDSWIFALAVLLCSTFVYNSMSTINHQALEQLHYVTELTELIKAKSSPRPDGVEDSTEFVSFFPDFLWTVRDFTLELKLNGHPITEDEYLENALKLIQGNNPRVQTSNFPRECIRRFFPKRKCFVFDRPTNDKDLLANIEKVSEKQLDPKFQEQTNIFCSYIFTHARTKTLREGITVTGNRLGTLAVTYVEAINSGAVPCLENAVITLAQRENSAAVQRASDYYSQQMAQRVKFPTDTLQELLDVHAACEREAIAIFMEHSFKDENQEFQKKFMETTMNKKGDFLLQNEESSVQYCQAKLNELSKGLMESISAGSFSVPGGHKLYMETKERIEQDYWQVPRKGVKAKEVFQRFLESQMVIEESILQSDKALTDREKAVAVDRAKKEAAEKEQELLKQKLQEQQQQMEAQVKSRKENIAQLKEKLQMEREHLLREQIMMLEHTQKVQNDWLHEGFKKKYEEMNAEISQFKRMIDTTKNDDTPWIARTLDNLADELTAILSAPAKLIGHGVKGVSSLFKKHKLPF | |
RUO | |
GBP6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |