missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GIT2 Partial ORF (NP_055591, 401 a.a. - 498 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009815-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121316

  • 6845.00 SEK / 25µg
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the GIT protein family, which interact with G protein-coupled receptor kinases and possess ADP-ribosylation factor (ARF) GTPase-activating protein (GAP) activity. GIT proteins traffic between cytoplasmic complexes, focal adhesions, and the cell periphery, and interact with Pak interacting exchange factor beta (PIX) to form large oligomeric complexes that transiently recruit other proteins. GIT proteins regulate cytoskeletal dynamics and participate in receptor internalization and membrane trafficking. This gene has been shown to repress lamellipodial extension and focal adhesion turnover, and is thought to regulate cell motility. This gene undergoes extensive alternative splicing to generate multiple isoforms, but the full-length nature of some of these variants has not been determined. The various isoforms have functional differences, with respect to ARF GAP activity and to G protein-coupled receptor kinase 2 binding. [provided by RefSeq]

Sequence: TDLETTASKTNRQKLQTLQSENSNLRKQATTNVYQVQTGSEYTDTSNHSSLKRRPSARGSRPMSMYETGSGQKPYLPMGEASRPEESRMRLQPFPAHA
Specifications

Specifications

NP_055591
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TDLETTASKTNRQKLQTLQSENSNLRKQATTNVYQVQTGSEYTDTSNHSSLKRRPSARGSRPMSMYETGSGQKPYLPMGEASRPEESRMRLQPFPAHA
RUO
GIT2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9815
GIT2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAT-2/DKFZp686G01261/KIAA0148/MGC760
GIT2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers