Learn More
Abnova™ Human GIT2 Partial ORF (NP_055591, 401 a.a. - 498 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4055.00 SEK - 6150.00 SEK
Specifications
Name | GIT2 (Human) Recombinant Protein (Q01) |
---|---|
Recombinant | Yes |
For Use With (Application) | Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) |
Molecular Weight | 36.52kDa |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16111316
|
Abnova™
H00009815-Q01.10UG |
10 ug |
4055.00 SEK
10µg |
Estimated Shipment: 06-10-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
16121316
|
Abnova™
H00009815-Q01.25UG |
25 ug |
6150.00 SEK
25µg |
Estimated Shipment: 06-10-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Description
This gene encodes a member of the GIT protein family, which interact with G protein-coupled receptor kinases and possess ADP-ribosylation factor (ARF) GTPase-activating protein (GAP) activity. GIT proteins traffic between cytoplasmic complexes, focal adhesions, and the cell periphery, and interact with Pak interacting exchange factor beta (PIX) to form large oligomeric complexes that transiently recruit other proteins. GIT proteins regulate cytoskeletal dynamics and participate in receptor internalization and membrane trafficking. This gene has been shown to repress lamellipodial extension and focal adhesion turnover, and is thought to regulate cell motility. This gene undergoes extensive alternative splicing to generate multiple isoforms, but the full-length nature of some of these variants has not been determined. The various isoforms have functional differences, with respect to ARF GAP activity and to G protein-coupled receptor kinase 2 binding. [provided by RefSeq]
Sequence: TDLETTASKTNRQKLQTLQSENSNLRKQATTNVYQVQTGSEYTDTSNHSSLKRRPSARGSRPMSMYETGSGQKPYLPMGEASRPEESRMRLQPFPAHASpecifications
GIT2 (Human) Recombinant Protein (Q01) | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
36.52kDa |