Learn More
Abnova™ Human GIT2 Partial ORF (NP_055591, 401 a.a. - 498 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009815-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the GIT protein family, which interact with G protein-coupled receptor kinases and possess ADP-ribosylation factor (ARF) GTPase-activating protein (GAP) activity. GIT proteins traffic between cytoplasmic complexes, focal adhesions, and the cell periphery, and interact with Pak interacting exchange factor beta (PIX) to form large oligomeric complexes that transiently recruit other proteins. GIT proteins regulate cytoskeletal dynamics and participate in receptor internalization and membrane trafficking. This gene has been shown to repress lamellipodial extension and focal adhesion turnover, and is thought to regulate cell motility. This gene undergoes extensive alternative splicing to generate multiple isoforms, but the full-length nature of some of these variants has not been determined. The various isoforms have functional differences, with respect to ARF GAP activity and to G protein-coupled receptor kinase 2 binding. [provided by RefSeq]
Sequence: TDLETTASKTNRQKLQTLQSENSNLRKQATTNVYQVQTGSEYTDTSNHSSLKRRPSARGSRPMSMYETGSGQKPYLPMGEASRPEESRMRLQPFPAHASpecifications
NP_055591 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TDLETTASKTNRQKLQTLQSENSNLRKQATTNVYQVQTGSEYTDTSNHSSLKRRPSARGSRPMSMYETGSGQKPYLPMGEASRPEESRMRLQPFPAHA | |
RUO | |
GIT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9815 | |
GIT2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAT-2/DKFZp686G01261/KIAA0148/MGC760 | |
GIT2 | |
Recombinant | |
wheat germ expression system |