missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GPR132 Partial ORF (NP_037477.1, 281 a.a. - 380 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
This gene encodes a subfamily member of the G-protein couple receptor (GPCR) superfamily. The encoded protein is a high-affinity receptor for lysophosphatidylcholine (LPC), a major phospholipid component of oxidized low density lipoprotein. This protein may react to LPC levels at sites of inflammation to limit the expansion of tissue-infiltrating cells. A similar protein in mouse is involved in cell cycle progression. [provided by RefSeq]
Sequence: GLEERLYTASVVFLCLSTVNGVADPIIYVLATDHSRQEVSRIHKGWKEWSMKTDVTRLTHSRDTEELQSPVALADHYTFSRPVHPPGSPCPAKRLIEESC
Specifications
Specifications
Accession Number | NP_037477.1 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 29933 |
Molecular Weight (g/mol) | 36.63kDa |
Name | GPR132 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue |
Quantity | 10 μg |
Immunogen | GLEERLYTASVVFLCLSTVNGVADPIIYVLATDHSRQEVSRIHKGWKEWSMKTDVTRLTHSRDTEELQSPVALADHYTFSRPVHPPGSPCPAKRLIEESC |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human GPR132 Partial ORF (NP_037477.1, 281 a.a. - 380 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction