missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human GUCY2F Partial ORF (NP_001513, 311 a.a. - 420 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16179084

Abnova™ Human GUCY2F Partial ORF (NP_001513, 311 a.a. - 420 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16179084
25 ug, 25µg
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16179084

Brand: Abnova™ H00002986Q01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a guanylyl cyclase found predominantly in photoreceptors in the retina. The encoded protein is thought to be involved in resynthesis of cGMP after light activation of the visual signal transduction cascade, allowing a return to the dark state. This protein is a single-pass type I membrane protein. Defects in this gene may be a cause of X-linked retinitis pigmentosa. [provided by RefSeq]

Sequence: VLTITVESQEKTFYQAFTEAAARGEIPEKLEFDQVSPLFGTIYNSIYFIAQAMNNAMKENGQAGAASLVQHSRNMQFHGFNQLMRTDSNGNGISEYVILDTNLKEWELHS

Specifications

Accession Number NP_001513
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2986
Molecular Weight (g/mol) 37.73kDa
Name GUCY2F (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen VLTITVESQEKTFYQAFTEAAARGEIPEKLEFDQVSPLFGTIYNSIYFIAQAMNNAMKENGQAGAASLVQHSRNMQFHGFNQLMRTDSNGNGISEYVILDTNLKEWELHS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CYGF/GC-F/GUC2DL/GUC2F/RETGC-2/ROS-GC2
Common Name GUCY2F
Gene Symbol GUCY2F
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human GUCY2F Partial ORF (NP_001513, 311 a.a. - 420 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.