missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HS6ST1 Partial ORF (NP_004798, 303 a.a. - 401 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009394-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16130546

  • 4515.00 SEK / 10µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the heparan sulfate biosynthetic enzyme family. Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biological activities. This enzyme is a type II integral membrane protein and is responsible for 6-O-sulfation of heparan sulfate. This enzyme does not share significant sequence similarity with other known sulfotransferases. A pseudogene located on chromosome 1 has been found for this gene. [provided by RefSeq]

Sequence: IRPFMQYNSTRAGGVEVDEDTIRRIEELNDLDMQLYDYAKDLFQQRYQYKRQLERREQRLRSREERLLHRAKEALPREDADEPGRVPTEDYMSHIIEKW
Specifications

Specifications

NP_004798
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IRPFMQYNSTRAGGVEVDEDTIRRIEELNDLDMQLYDYAKDLFQQRYQYKRQLERREQRLRSREERLLHRAKEALPREDADEPGRVPTEDYMSHIIEKW
RUO
HS6ST1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9394
HS6ST1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp547H098/FLJ25392/HS6ST/MGC116899/MGC116901
HS6ST1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers