missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human IFN-beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB11127-100
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IFN-beta Monoclonal antibody specifically detects IFN-beta in Human samples. It is validated for Neutralization
Specifikationer
| IFN-beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| CHO-expressed recombinant Human IFN-beta protein, Met22-Asn187, Accession # P01574 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization | |
| 1020369 | |
| Neutralization | |
| P01574 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3456 | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering