missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IGHA1 Full-length ORF (AAH05951, 1 a.a. - 500 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003493-P02.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MDWTWSILFLVAAATGAQSQVHLVQSGAEVMSPGASVRVSCKTSGYAFHTYSIIWVRQAPGQGLEWMGWISPSSDNTRFAKKFQGRVTLTTDTSTSTVYMELRSLRSDDTAVYYCARRYCSYSSCQNDYYYYYMDVWGKGTTVTVSSASPTSPKVFPLSLCSTQPDGNVVIACLVQGFFPQEPLSVTWSESGQGVTARNFPPSQDASGDLYTTSSQLTLPATQCLAGKSVTCHVKHYTNPSQDVTVPCPVPSTPPTPSPSTPPTPSPSCCHPRLSLHRPALEDLLLGSEANLTCTLTGLRDASGVTFTWTPSSGKSAVQGPPDRDLCGCYSVSSVLSGCAEPWNHGKTFTCTAAYPESKTPLTATLSKSGNTFRPEVHLLPPPSEELALNELVTLTCLARGFSPKDVLVRWLQGSQELPREKYLTWASRQEPSQGTTTFAVTSILRVAAEDWKKGDTFSCMVGHEALPLAFTQETIDRLAGKPTHVNVSVVMAEVDGTCYSpecifications
AAH05951 | |
Liquid | |
3493 | |
IGHA1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDWTWSILFLVAAATGAQSQVHLVQSGAEVMSPGASVRVSCKTSGYAFHTYSIIWVRQAPGQGLEWMGWISPSSDNTRFAKKFQGRVTLTTDTSTSTVYMELRSLRSDDTAVYYCARRYCSYSSCQNDYYYYYMDVWGKGTTVTVSSASPTSPKVFPLSLCSTQPDGNVVIACLVQGFFPQEPLSVTWSESGQGVTARNFPPSQDASGDLYTTSSQLTLPATQCLAGKSVTCHVKHYTNPSQDVTVPCPVPSTPPTPSPSTPPTPSPSCCHPRLSLHRPALEDLLLGSEANLTCTLTGLRDASGVTFTWTPSSGKSAVQGPPDRDLCGCYSVSSVLSGCAEPWNHGKTFTCTAAYPESKTPLTATLSKSGNTFRPEVHLLPPPSEELALNELVTLTCLARGFSPKDVLVRWLQGSQELPREKYLTWASRQEPSQGTTTFAVTSILRVAAEDWKKGDTFSCMVGHEALPLAFTQETIDRLAGKPTHVNVSVVMAEVDGTCY | |
RUO | |
IGHA1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
80.74kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ14473/FLJ35065/FLJ35500/FLJ36402/FLJ39698/FLJ40001/FLJ41548/FLJ41552/FLJ41789/FLJ43248/FLJ43594/FLJ44293/FLJ46028/FLJ46621/FLJ46724/FLJ46811/FLJ46824/IgA1/MGC102857 | |
IGHA1 | |
Yes | |
wheat germ expression system |