Läs mer
Human IL-17RB PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 3 publications
Brand:  R&D Systems FAB1207P-025
 Se fler versioner av denna produkt
Produktkod. 18733658
  • 2915.00 SEK / 25 tester
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Beskrivning
Beskrivning
IL-17RB Monoclonal specifically detects IL-17RB in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifikationer
Specifikationer
IL-17RB
Monoclonal
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CRL4IL-17 receptor homolog 1, cytokine receptor CRL4, Cytokine receptor-like 4, Evi27, EVI27IL-17B receptor, IL-17 RB, IL-17 receptor B, IL-17 RH1, IL17BRinterleukin 17B receptor, IL-17ER, IL17RB, IL-17RB, IL17Rh1, IL-17Rh1, IL17RH1MGC5245, interleukin 17 receptor B, interleukin 17 receptor homolog 1, interleukin-17 receptor B, Interleukin-17B receptor
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
55540
Human
Purified
Flow Cytometry
170220
Flow Cytometry 10 uL/10^6 cells
Q9NRM6
IL17RB
Mouse myeloma cell line NS0-derived recombinant human IL-17 RB Arg18-Gly289 Accession # Q9NRM6
25 Tests
Primary
Detects human IL-17 RB in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse (rm) IL-17 RB, recombinant human IL-17 R, or rmIL-17 R.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2b
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Produktspecifikationer
Kampanjer
Kampanjer