missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human IL-17RB PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems FAB1207P-100
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IL-17RB Monoclonal specifically detects IL-17RB in Human samples. It is validated for Flow Cytometry.
Specifikationer
| IL-17RB | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CRL4IL-17 receptor homolog 1, cytokine receptor CRL4, Cytokine receptor-like 4, Evi27, EVI27IL-17B receptor, IL-17 RB, IL-17 receptor B, IL-17 RH1, IL17BRinterleukin 17B receptor, IL-17ER, IL17RB, IL-17RB, IL17Rh1, IL-17Rh1, IL17RH1MGC5245, interleukin 17 receptor B, interleukin 17 receptor homolog 1, interleukin-17 receptor B, Interleukin-17B receptor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 55540 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 170220 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q9NRM6 | |
| IL17RB | |
| Mouse myeloma cell line NS0-derived recombinant human IL-17 RB Arg18-Gly289 Accession # Q9NRM6 | |
| 100 Tests | |
| Primary | |
| Detects human IL-17 RB in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse (rm) IL-17 RB, recombinant human IL-17 R, or rmIL-17 R. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering