missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human IL-2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB104421-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IL-2 Monoclonal antibody specifically detects IL-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry
Specifikationer
| IL-2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF | |
| E. coli-derived human IL-2, Ala21-Thr153, Accession # P60568 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry | |
| 1019340 | |
| Western Blot 2.5 ug/mL, ELISA, Immunocytochemistry 8-25 ug/mL | |
| P60568 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3558 | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering