missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human JAK3 (NP_000206.1, 781 a.a. - 1124 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells Product Code.: 16047926

Abnova™ Human JAK3 (NP_000206.1, 781 a.a. - 1124 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells

Product Code. 16047926
100μg
Click to view available options
:
100μg
This item is not returnable. View return policy

Product Code. 16047926

Brand: Abnova™ P4709.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

The protein encoded by this gene is a member of the Janus kinase (JAK) family of tyrosine kinases involved in cytokine receptor-mediated intracellular signal transduction. It is predominantly expressed in immune cells and transduces a signal in response to its activation via tyrosine phosphorylation by interleukin receptors. Mutations in this gene are associated with autosomal SCID (severe combined immunodeficiency disease). [provided by RefSeq]

Specifications

Accession Number NP_000206.1
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 3718
Molecular Weight (g/mol) 68.379kDa
Name JAK3 (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Purification Method GST-Affinity Chromatography
Quality Control Testing 2 ug/lane SDS-PAGE Stained with Coomassie Blue
Quantity 100 ug
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing
Regulatory Status RUO
Gene Alias JAK-3/JAK3_HUMAN/JAKL/L-JAK/LJAK
Common Name JAK3
Gene Symbol JAK3
Biological Activity 5pmol/μg x min
Species Baculovirus
Recombinant Recombinant
Protein Tag GST-His
Expression System Insect cell (Sf9) expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human JAK3 (NP_000206.1, 781 a.a. - 1124 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.