missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human Kir2Ds5 Full-Length Orf (Aai60061.1, 1 A.A.-304 A.A.) Recombinant Protein With Gst Tag At N-Terminal.
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: MSLMVISMACVAFFLLQGAWPHEGFRRKPSLLAHPGPLVKSEETVILQCWSDVMFEHFLLHREGTFNHTLRLIGEHIDGVSKGNFSIGRMTQDLAGTYRCYGSVTHSPYQLSAPSDPLDIVITGLYEKPSLSAQPGPTVLAGESVT
Specifications
Specifications
Accession Number | AAI60061.1 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3810 |
Name | KIR2DS5 (Human) Recombinant Protein (P01) |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue |
Quantity | 25 μg |
Immunogen | MSLMVISMACVAFFLLQGAWPHEGFRRKPSLLAHPGPLVKSEETVILQCWSDVMFEHFLLHREGTFNHTLRLIGEHIDGVSKGNFSIGRMTQDLAGTYRCYGSVTHSPYQLSAPSDPLDIVITGLYEKPSLSAQPGPTVLAGESVTLSCSSRSSYDMYHLSREGEAHERRLPAGPKVNRTFQADFPLDPATHGGTYRCFGSFRDSPYEWSKSSDPLLVSVTGNSSNSWPSPTEPSSETGNPRHLHVLIGTSVVKLPFTILLFFLLHRWCSNKKNASVMDQGPAGNRTVNREDSDEQDHQEVSYA |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human Kir2Ds5 Full-Length Orf (Aai60061.1, 1 A.A.-304 A.A.) Recombinant Protein With Gst Tag At N-Terminal. >
Spot an opportunity for improvement?Share a Content Correction