missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human KLC3 Full-length ORF (AAH73841.1, 1 a.a. - 503 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16178473

Abnova™ Human KLC3 Full-length ORF (AAH73841.1, 1 a.a. - 503 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16178473
10 μg, 10µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16178473

Brand: Abnova™ H00147700P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the kinesin light chain gene family. Kinesins are molecular motors involved in the transport of cargo along microtubules, and are composed of two kinesin heavy chain (KHC) and two kinesin light chain (KLC) molecules. KLCs are thought to typically be involved in binding cargo and regulating kinesin activity. In the rat, a protein similar to this gene product is expressed in post-meiotic spermatids, where it associates with structural components of sperm tails and mitochondria. [provided by RefSeq]

Sequence: MSVQVAAPGSAGLGPERLSPEELVRQTRQVVQGLEALRAEHHGLAGHLAEALAGQGPAAGLEMLEEKQQVVSHSLEAIELGLGEAQVLLALSAHVGALEAEKQRLRSQARRLAQENVWLREELEETQRRLRASEESVAQLEEEKRHLEFLGQLRQYDPPAESQSESPPRRDSLASLFPSEEEERKGPEAAGAAAAQQGGYEIPARLRTLHNLVIQYAGQGRYEVAVPLCRQALEDLERSSGHCHPDVATMLNILALVYRDQNKYKEATDLLHDALQIREQTLGPEHPAVAATLNNLAVLYGKRGRYREAEPLCQRALEIREKVLGADHPDVAKQLNNLALLCQNQGKFEDVERHYARALSIYEALGGPHDPNVAKTKNNLASAYLKQNKYQQAEELYKEILHKEDLPAPLGAPNTGTAGDAEQALRRSSSLSKIRESIRRGSEKLVSRLRGEAAAGAAGMKRAMSLNTLNVDAPRAPGTQFPSWHLDKAPRTLSASTQDLSPH

Specifications

Accession Number AAH73841.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 147700
Molecular Weight (g/mol) 81.6kDa
Name KLC3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MSVQVAAPGSAGLGPERLSPEELVRQTRQVVQGLEALRAEHHGLAGHLAEALAGQGPAAGLEMLEEKQQVVSHSLEAIELGLGEAQVLLALSAHVGALEAEKQRLRSQARRLAQENVWLREELEETQRRLRASEESVAQLEEEKRHLEFLGQLRQYDPPAESQSESPPRRDSLASLFPSEEEERKGPEAAGAAAAQQGGYEIPARLRTLHNLVIQYAGQGRYEVAVPLCRQALEDLERSSGHCHPDVATMLNILALVYRDQNKYKEATDLLHDALQIREQTLGPEHPAVAATLNNLAVLYGKRGRYREAEPLCQRALEIREKVLGADHPDVAKQLNNLALLCQNQGKFEDVERHYARALSIYEALGGPHDPNVAKTKNNLASAYLKQNKYQQAEELYKEILHKEDLPAPLGAPNTGTAGDAEQALRRSSSLSKIRESIRRGSEKLVSRLRGEAAAGAAGMKRAMSLNTLNVDAPRAPGTQFPSWHLDKAPRTLSASTQDLSPH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KLC2/KLC2L/KLCt/KNS2B
Common Name KLC3
Gene Symbol KLC3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human KLC3 Full-length ORF (AAH73841.1, 1 a.a. - 503 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.