Learn More
Abnova™ Human KREMEN2 Full-length ORF (NP_078783.1, 1 a.a. - 420 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079412-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. The encoded protein forms a ternary membrane complex with DKK1 and the WNT receptor lipoprotein receptor-related protein 6 (LRP6), and induces rapid endocytosis and removal of LRP6 from the plasma membrane. It contains extracellular kringle, WSC, and CUB domains. Alternatively spliced transcript variants encoding distinct isoforms have been observed for this gene. [provided by RefSeq]
Sequence: MGTQALQGFLFLLFLPLLQPRGASAGSLHSPGLSECFQVNGADYRGHQNRTGPRGAGRPCLFWDQTQQHSYSSASDPHGRWGLGAHNFCRNPDGDVQPWCYVAETEEGIYWRYCDIPSCHMPGYLGCFVDSGAPPALSGPSGTSTKLTVQVCLRFCRMKGYQLAGVEAGYACFCGSESDLARGRLAPATDCDQICFGHPGQLCGGDGRLGVYEVSVGSCQGNWTAPQGVIYSPDFPDEYGPDRNCSWALGPPGAALELTFRLFELADPRDRLELRDAASGSLLRAFDGARPPPSGPLRLGTAALLLTFRSDARGHAQGFALTYRGLQDAAEDPEAPEGSAQTPAAPLDGANVSCSPRPGAPPAAIGGAVCWLREKGPRRWGLPGAPGEAGLCGTNSPEGWPCPAPPGTPRLRVLPRATGLSpecifications
NP_078783.1 | |
Liquid | |
79412 | |
KREMEN2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGTQALQGFLFLLFLPLLQPRGASAGSLHSPGLSECFQVNGADYRGHQNRTGPRGAGRPCLFWDQTQQHSYSSASDPHGRWGLGAHNFCRNPDGDVQPWCYVAETEEGIYWRYCDIPSCHMPGYLGCFVDSGAPPALSGPSGTSTKLTVQVCLRFCRMKGYQLAGVEAGYACFCGSESDLARGRLAPATDCDQICFGHPGQLCGGDGRLGVYEVSVGSCQGNWTAPQGVIYSPDFPDEYGPDRNCSWALGPPGAALELTFRLFELADPRDRLELRDAASGSLLRAFDGARPPPSGPLRLGTAALLLTFRSDARGHAQGFALTYRGLQDAAEDPEAPEGSAQTPAAPLDGANVSCSPRPGAPPAAIGGAVCWLREKGPRRWGLPGAPGEAGLCGTNSPEGWPCPAPPGTPRLRVLPRATGL | |
RUO | |
KREMEN2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KRM2/MGC10791/MGC16709 | |
KREMEN2 | |
Yes | |
wheat germ expression system |