missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human LBP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB870R-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
LBP Monoclonal specifically detects LBP in Human samples. It is validated for ELISA.
Specifikationer
| LBP | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| lipopolysaccharide binding protein, lipopolysaccharide-binding protein, LPS-binding protein, MGC22233 | |
| Mouse myeloma cell line NS0-derived recombinant human LBP Ala26-Val481 Accession # AAD21962 | |
| 25 μg | |
| 3929 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 127408R | |
| ELISA | |
| AAD21962 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human LBP in direct ELISAs. | |
| Human | |
| Supernatant |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering