Learn More
Abnova™ Human LDHAL6B Partial ORF (NP_005497, 282 a.a. - 381 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16131227
Abnova™ Human LDHAL6B Partial ORF (NP_005497, 282 a.a. - 381 a.a.) Recombinant Protein with GST-tag at N-terminal
Product Code. 16131227
10 μg, 10µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16131227
Brand: Abnova™ H00092483Q01.10ug
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used for AP, Array, ELISA, WB-Re
Sequence: EVTATAYEIIKMKGYTSWAIGLSVADLTESILKNLRRIHPVSTITKGLYGIDEEVFLSIPCILGENGITNLIKIKLTPEEEAHLKKSAKTLWEIQNKLKL
Specifications
Accession Number NP_005497
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 92483
Molecular Weight (g/mol) 36.74kDa
Name LDHAL6B (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen EVTATAYEIIKMKGYTSWAIGLSVADLTESILKNLRRIHPVSTITKGLYGIDEEVFLSIPCILGENGITNLIKIKLTPEEEAHLKKSAKTLWEIQNKLKL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias LDHAL6/LDHL
Common Name LDHAL6B
Gene Symbol LDHAL6B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction
Product Title
Abnova™ Human LDHAL6B Partial ORF (NP_005497, 282 a.a. - 381 a.a.) Recombinant Protein with GST-tag at N-terminal >