missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MARCH5 Partial ORF (NP_060294, 1 a.a. - 95 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
MARCH5 is a ubiquitin ligase of the mitochondrial outer membrane that plays a role in the control of mitochondrial morphology by regulating mitofusin-2 (MFN2; MIM 608507) and DRP1 (DNM1L; MIM 603850) (Nakamura et al., 2006 [PubMed 16936636]).[supplied by OMIM]
Sequence: MPDQALQQMLDRSCWVCFATDEDDRTAEWVRPCRCRGSTKWVHQACLQRWVDEKQRGNSTARVACPQCNAEYLIVFPKLGPVVYVLDLADRLISK
Specifications
Specifications
Accession Number | NP_060294 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 54708 |
Molecular Weight (g/mol) | 36.19kDa |
Name | MARCH5 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | MPDQALQQMLDRSCWVCFATDEDDRTAEWVRPCRCRGSTKWVHQACLQRWVDEKQRGNSTARVACPQCNAEYLIVFPKLGPVVYVLDLADRLISK |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Human MARCH5 Partial ORF (NP_060294, 1 a.a. - 95 a.a.) Recombinant Protein with GST-tag at N-terminal >
Spot an opportunity for improvement?Share a Content Correction