missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MC3R Partial ORF (NP_063941, 1 a.a. - 74 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004159-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16177641

  • 4515.00 SEK / 10µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a G-protein-coupled receptor for melanocyte-stimulating hormone and adrenocorticotropic hormone that is expressed in tissues other than the adrenal cortex and melanocytes. This gene maps to the same region as the locus for benign neonatal epilepsy. Mice deficient for this gene have increased fat mass despite decreased food intake, suggesting a role for this gene product in the regulation of energy homeostasis. Mutations in this gene are associated with a susceptibility to obesity in humans. [provided by RefSeq]

Sequence: MSIQKTYLEGDFVFPVSSSSFLRTLLEPQLGSALLTAMNASCCLPSVQPTLPNGSEHLQAPFFSNQSSSAFCEQ
Specifications

Specifications

NP_063941
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.88kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MSIQKTYLEGDFVFPVSSSSFLRTLLEPQLGSALLTAMNASCCLPSVQPTLPNGSEHLQAPFFSNQSSSAFCEQ
RUO
MC3R
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4159
MC3R (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BMIQ9/MC3/MC3-R
MC3R
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers