missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MC3R Partial ORF (NP_063941, 1 a.a. - 74 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_063941 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4159 |
Molecular Weight (g/mol) | 33.88kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16187641
|
Abnova™
H00004159-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16177641
|
Abnova™
H00004159-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a G-protein-coupled receptor for melanocyte-stimulating hormone and adrenocorticotropic hormone that is expressed in tissues other than the adrenal cortex and melanocytes. This gene maps to the same region as the locus for benign neonatal epilepsy. Mice deficient for this gene have increased fat mass despite decreased food intake, suggesting a role for this gene product in the regulation of energy homeostasis. Mutations in this gene are associated with a susceptibility to obesity in humans. [provided by RefSeq]
Sequence: MSIQKTYLEGDFVFPVSSSSFLRTLLEPQLGSALLTAMNASCCLPSVQPTLPNGSEHLQAPFFSNQSSSAFCEQSpecifications
NP_063941 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.88kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BMIQ9/MC3/MC3-R | |
MC3R | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4159 | |
MC3R (Human) Recombinant Protein (Q01) | |
MSIQKTYLEGDFVFPVSSSSFLRTLLEPQLGSALLTAMNASCCLPSVQPTLPNGSEHLQAPFFSNQSSSAFCEQ | |
RUO | |
MC3R | |
Wheat Germ (in vitro) | |
GST | |
Liquid |