missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human MED22 Full-length ORF (NP_852468.1, 1 a.a. - 140 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16181152

Abnova™ Human MED22 Full-length ORF (NP_852468.1, 1 a.a. - 140 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16181152
25 ug, 25µg
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16181152

Brand: Abnova™ H00006837P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene is located in the surfeit gene cluster, a group of very tightly linked housekeeping genes that do not share sequence similarity. The gene is oriented in a head-to-head fashion with RPL7A (SURF3) and the two genes share a bidirectional promoter. The encoded proteins are localized to the cytoplasm. Two alternative transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MAQQRALPQSKETLLQSYNKRLKDDIKSIMDNFTEIIKTAKIEDETQVSRATQGEQDNYEMHVRAANIVRAGESLMKLVSDLKQFLILNDFPSVNEAIDQRNQQLRTLQEECDRKLITLRDEISIDLYELEEEYYSSRYK

Specifications

Accession Number NP_852468.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6837
Molecular Weight (g/mol) 42.9kDa
Name MED22 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MAQQRALPQSKETLLQSYNKRLKDDIKSIMDNFTEIIKTAKIEDETQVSRATQGEQDNYEMHVRAANIVRAGESLMKLVSDLKQFLILNDFPSVNEAIDQRNQQLRTLQEECDRKLITLRDEISIDLYELEEEYYSSRYK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MED24/MGC48682/SURF5
Common Name MED22
Gene Symbol MED22
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human MED22 Full-length ORF (NP_852468.1, 1 a.a. - 140 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.