missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse HIF-1 alpha/HIF1A Alexa Fluor 488-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC1935G-100UG
This item is not returnable.
View return policy
Description
HIF-1 alpha Monoclonal specifically detects HIF-1 alpha in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
HIF-1 alpha | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q16665.1 | |
HIF1A | |
E. coli-derived recombinant human HIF-1 alpha Arg575-Asn826 Accession # Q16665.1 | |
100 μg | |
Primary | |
Recognizes human and mouse HIF-1 alpha in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry | |
241812 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF 1A, HIF1, HIF1 alpha, HIF-1 alpha, HIF-1a, HIF1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3091 | |
Human, Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human/Mouse HIF-1 alpha/HIF1A Alexa Fluor 488-conjugated Antibody, R&D Systems™ > 100ug; Alexa Fluor 488
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction