Learn More
Abnova™ Human MSX2 Partial ORF (AAH15509.1, 1 a.a. - 133 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This gene encodes a member of the muscle segment homeobox gene family. The encoded protein is a transcriptional repressor whose normal activity may establish a balance between survival and apoptosis of neural crest-derived cells required for proper craniofacial morphogenesis. The encoded protein may also have a role in promoting cell growth under certain conditions and may be an important target for the RAS signaling pathways. Mutations in this gene are associated with parietal foramina 1 and craniosynostosis type 2. [provided by RefSeq]
Specifications
Specifications
Accession Number | AAH15509.1 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4488 |
Molecular Weight (g/mol) | 40.26kDa |
Name | MSX2 (Human) Recombinant Protein (Q03) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue |
Quantity | 25 ug |
Immunogen | MASPSKGNDLFSPDEEGPAVVAGPGPGPGGAEGAAEERRVKVSSLPFSVEALMSDKKPPKEASPLPAESASAGATLRPLLLSGHGAREAHSPGPLVKPFETASVKSENSEDGAAWMQEPGRYSPPPRHTSPTT |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.