Learn More
Abnova™ Human MYL5 Full-length ORF (AAH40050.1, 1 a.a. - 132 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004636-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes one of the myosin light chains, a component of the hexameric ATPase cellular motor protein myosin. Myosin is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene product, one of the regulatory light chains, is expressed in fetal muscle and in adult retina, cerebellum, and basal ganglia. [provided by RefSeq]
Sequence: MDQNRDGFIDKEDLKDTYASLGKTNVKDDELDAMLKEASGPINFTMFLNLFGEKLSGTDAEETILNAFKMLDPDGKGKINKEYIKRLLMSQADKMTAEEVDQMFQFASIDVAGNLDYKALSYVITHGEEKEESpecifications
AAH40050.1 | |
Liquid | |
4636 | |
Human MYL5 Full-length ORF (AAH40050.1, 1 a.a. - 132 a.a.) Recombinant Protein with GST-tag at N-terminal | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at −80°C. Aliquot to avoid repeated freezing and thawing. | |
MYL5 | |
Wheat Germ (in vitro) | |
GST Tag |
AP, Array, ELISA, WB-Re | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.26kDa | |
Glutathione Sepharose 4 Fast Flow | |
25μg | |
RUO | |
MYL5 | |
Yes | |
wheat germ expression system |