missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human NKX2-3 Partial ORF (NP_660328.1, 1 a.a. - 56 a.a.) Recombinant Protein with GST-tag at N-terminal Product Code.: 16132237

Abnova™ Human NKX2-3 Partial ORF (NP_660328.1, 1 a.a. - 56 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16132237
25 μg, 25µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16132237

Brand: Abnova™ H00159296Q01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

NKX2C is a member of the NKX family of homeodomain-containing transcription factors, which are implicated in many aspects of cell type specification and maintenance of differentiated tissue functions. See Harvey (1996) [PubMed 8812123] for a review of the structure, regulation, function, and evolution of NK2 homeobox genes with an emphasis on their roles in heart development.[supplied by OMIM]

Sequence: MMLPSPVTSTPFSVKDILNLEQQHQHFHGAHLQADLEHHFHSAPCMLAAAEGTQFS

Specifications

Accession Number NP_660328.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 159296
Molecular Weight (g/mol) 31.9kDa
Name NKX2-3 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MMLPSPVTSTPFSVKDILNLEQQHQHFHGAHLQADLEHHFHSAPCMLAAAEGTQFS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CSX3/NK2.3/NKX2.3/NKX2C/NKX4-3
Common Name NKX2-3
Gene Symbol NKX2-3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human NKX2-3 Partial ORF (NP_660328.1, 1 a.a. - 56 a.a.) Recombinant Protein with GST-tag at N-terminal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.