Learn More
Abnova™ Human NOTCH2 Partial ORF (NP_077719, 27 a.a. - 136 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This gene encodes a member of the Notch family. Members of this Type 1 transmembrane protein family share structural characteristics including an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. Notch family members play a role in a variety of developmental processes by controlling cell fate decisions. The Notch signaling network is an evolutionarily conserved intercellular signaling pathway which regulates interactions between physically adjacent cells. In Drosophilia, notch interaction with its cell-bound ligands (delta, serrate) establishes an intercellular signaling pathway that plays a key role in development. Homologues of the notch-ligands have also been identified in human, but precise interactions between these ligands and the human notch homologues remain to be determined. This protein is cleaved in the trans-Golgi network, and presented on the cell surface as a heterodimer. This protein functions as a receptor for membrane bound ligands, and may play a role in vascular, renal and hepatic development. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_077719 |
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4853 |
Molecular Weight (g/mol) | 37.84kDa |
Name | NOTCH2 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 ug |
Immunogen | QCRDGYEPCVNEGMCVTYHNGTGYCKCPEGFLGEYCQHRDPCEKNRCQNGGTCVAQAMLGKATCRCASGFTGEDCQYSTSHPCFVSRPCLNGGTCHMLSRDTYECTCQVG |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.