Learn More
Abnova™ Human OR8B4 Full-length ORF (NP_001005196.1, 1 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]
Specifications
Specifications
Accession Number | NP_001005196.1 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 283162 |
Molecular Weight (g/mol) | 60.8kDa |
Name | OR8B4 (Human) Recombinant Protein (P01) |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 μg |
Immunogen | MTLRNSSSVTEFILVGLSEQPELQLPLFLLFLGIYVFTVVGNLGLITLIGINPSLHTPMYFFLFNLSFIDLCYSCVFTPKMLNDFVSESIISYVGCMTQLFFFCFFVNSECYVLVSMAYDRYVAICNPLLYMVTMSPRVCFLLMFGSYVVGFAGAMAHTGSMLRLTFCDSNVIDHYLCDVLPLLQLSCTSTHVSELVFFIVVGVITMLSSISIVISYALILSNILCIPSAEGRSKAFSTWGSHIIAVALFFGSGTFTYLTTSFPGSMNHGRFASVFYTNVVPMLNPSIYSLRNKDDKLALGKTLKRVLF |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.